Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CYLD Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
PW0760-0100 PW0760-0025
89161-948EA 500 USD InStock
89161-948 89161-946
Anti-CYLD Rabbit Polyclonal Antibody
Antibodies
The tumor suppressor CYLD is a deubiquitinylating enzyme (DUB) that antagonises NF-κB and JNK signaling by disassembly of Lys63-linked ubiquitin chains synthesised in response to cytokine stimulation. The DUB activity of CYLD is mediated by its C-terminal ubiquitin specific protease (USP) domain, pathogenic truncations and mutations of which are associated with the hypertrophic skin tumor cylindromatosis through disruption of the USP domain and subsequent loss of CYLD catalytic activity.

  • For Western Blot

Recognizes human CYLD.

Type: Primary
Antigen: CYLD
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty