Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Ubiquitin Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
PW0930-0100 PW0930-1000
89161-956EA 1410 USD InStock
89161-956 89161-954
Anti-Ubiquitin Mouse Monoclonal Antibody
Antibodies
Ubiquitin modification of cellular proteins has been shown to be involved with a wide range of biochemical processes including proteasomal degradation, signal transduction, DNA repair, endocytosis and autophagy.

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes mono- and poly-ubiquitin protein conjugates, free polyubiquitin chains and free ubiquitin.

Type: Primary
Antigen: UBIQUITIN
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Species Independent

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty