Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PSMA6 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
PW8100-0100 PW8100-0025
89161-980EA 175 USD InStock
89161-980 89161-982
Anti-PSMA6 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human and rabbit proteasome 20S (alpha6 subunit). Does not cross-react with rat or mouse protein.

Type: Primary
Antigen: PSMA6
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty