Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Proteasome 11S Alpha Subunit Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
PW8185-0025 PW8185-0100
89162-044EA 212 USD InStock
89162-044 89162-046
Anti-Proteasome 11S Alpha Subunit Rabbit Polyclonal Antibody
Antibodies
The proteasome is widely recognised as the central enzyme of non-lysosomal protein degradation. It is responsible for intracellular protein turnover and it is also critically involved in many regulatory processes and, in higher eukaryotes, in antigen processing. An enzymatic cascade is responsible for the attachment of multiple ubiquitin molecules to lysine residues of proteins targeted for degradation. The 26S proteasome is the key enzyme of the ubiquitin/ATP-dependent pathway of protein degradation.

The catalytic core of this unusually large (2000kDa, 450Å in length) complex (31 subunits) is formed by the 20S proteasome, a barrel shaped structure shown by electron microscopy to comprise of four rings each containing seven subunits. 20S Proteasomes degrade only unfolded proteins in an energy-independent manner, whereas 26S proteasomes degrade native and ubiquitinylated proteins in an ATP-dependent manner. The native protein substrates are recognised by subunits, some with ATP binding sites, of the outer 19S caps of the 26S proteasome. The 19S regulatory subunit recognizes ubiquitinated proteins and plays an essential role in unfolding and translocating targets into the lumen of the 20S subunit. A second activator which can associate with the 20S proteasome in the absence of ATP is known as PA28 or the 11S regulator. The pure PA28 activator is a complex of two alternating subunits, PA28α and PA28β, which share approximately 50% homology but also show considerable similarity (30-40%) to a nuclear protein of unknown function, the Ki autoantigen (recently referred to as PA28γ). These subunits, with an apparent relative molecular weight of approximately 29kDa, form ringlike heteromeric complexes of ~200kDa possibly with an α3β3 stoichiometry. Electron microscopic studies have shown PA28 to be a ring shaped particle which, like the 19S, caps the 20S proteasome, by binding to the α-rings, at both or either end. The complex may, however, be readily dissociated. The finding that PA28 modulates the proteasome-catalysed production of antigenic peptides presented to the immune system on MHC class I molecules indicates a cellular function of this activator in antigen processing. Several genetic diseases are associated with defects in the ubiquitin-proteasome pathway. Some examples of affected proteins include those linked to cystic fibrosis, Angelman's syndrome, and Liddle syndrome.

  • For Western Blot

Recognizes human, mouse and rat proteasome activator 11S (alpha subunit).

Type: Primary
Antigen: PROTEASOME 11S ALPHA SUBUNIT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty