Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Proteasome 19S Rpn7/S10A Subunit Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
PW8225-0100 PW8225-0025
89162-076EA 200 USD InStock
89162-076 89162-078
Anti-Proteasome 19S Rpn7/S10A Subunit Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunoprecipitation, Western Blot

Recognizes human and yeast proteasome 26S (19S regulator complex) (Rpn7/S10a subunit).

Type: Primary
Antigen: PROTEASOME 19S RPN7/S10A SUBUNIT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty