Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Proteasome 19S Rpt1/S7 Subunit Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
PW8255-0025 PW8255-0100
89162-100EA 154 USD InStock
89162-100 89162-102
Anti-Proteasome 19S Rpt1/S7 Subunit Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Western Blot

Recognizes yeast proteasome 26S (19S regulator complex) (Rpt1/S7 subunit).

Type: Primary
Antigen: PROTEASOME 19S RPT1/S7 SUBUNIT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Yeast

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty