Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PSMA5 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
PW8270-0100
89162-110EA 534 USD InStock
89162-110
Anti-PSMA5 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG

  • For Immunohistochemistry, Immunoprecipitation

Recognizes human proteasome 20S (alpha5 subunit).

Type: Primary
Antigen: PSMA5
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty