Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Cop9 Signalosome Csn6 Subunit Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
PW8295-0025 PW8295-0100
89162-124EA 154 USD InStock
89162-124 89162-126
Anti-Cop9 Signalosome Csn6 Subunit Rabbit Polyclonal Antibody
Antibodies
The polyclonal antibody to Csn6 was generated by immunisation of rabbits with a synthetic peptide corresponding to residues [283-297] of the human Csn6 sequence. The antibody has been characterised by one-dimensional Western blotting. Vial contains a partially purified immunoglobulin preparation suspended phosphate-buffered saline containing 0.01M sodium azide.  The COP9 signalosome, once defined as a repressor complex of light activated development in Arabidopsis, has recently been found in humans and is probably present in most multicellular organisms. The COP9 signalosome is closely related to the lid sub-complex of the 26S proteasome in structural composition and probably shares a common evolutionary ancestor. A multifaceted role of the COP9 signalosome in cell-signalling processes is hinted at by its associated novel kinase activity, as well as the involvement of its subunits in regulating multiple cell-signalling pathways and cell-cycle progression. The molecular genetic studies in Arabidopsis suggest that the complex functions as part of a highly conserved regulatory network, whose physiological rüle in animals has yet to be determined. The COP9 signalosome, also known as the the COP9 complex and JAB1-containing signalosome, is a conserved nuclear protein complex found in plants and animals. It is composed of eight distinct subunits, designated S1 to S8 (very recently re-designated Csn1-Csn8) according to the molecular weight of the mammalian complex subunits. Subunit composition and the subunit sequences are substantially conserved between the mammalian and the plant complexes, implying that the complex has a conserved cellular function in higher eukaryotic organisms. The role and function of Csn6 has yet to be determined although this subunit is known to interact with subunits Csn2 and Csn7 within the COP9 signalosome.

  • For Western Blot

Recognizes human COP9 signalosome (Csn6 subunit).

Type: Primary
Antigen: COP9 SIGNALOSOME CSN6 SUBUNIT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty