Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Cop9 Signalosome Csn7 Subunit Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
PW8300-0025 PW8300-0100
89162-130EA 534 USD InStock
89162-130 89162-128
Anti-Cop9 Signalosome Csn7 Subunit Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunoprecipitation, Western Blot

Recognizes human COP9 signalosome (Csn7 subunit).

Type: Primary
Antigen: COP9 SIGNALOSOME CSN7 SUBUNIT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty