Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Proteasome Activator 11S Subunit Antibody Sampler Pack

Supplier: Enzo Life Sciences
PW8915-0001
89162-272EA 250 USD InStock
89162-272
Proteasome Activator 11S Subunit Antibody Sampler Pack
Antibodies
Contains 10 µl each of BML-PW8185, BML-PW8190, and BML-PW8240.

Type:
Antigen: PSME11
Clonality:
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty