Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PSMB2 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
PW9300-0100 PW9300-0025
89162-324EA 174.47 USD InStock
89162-324 89162-326
Anti-PSMB2 Mouse Monoclonal Antibody
Antibodies
The hybridoma secreting the antibody to subunit β2 was generated by fusion of splenocytes from Balb/c mice which had received repeated immunization with dinitrophenylated proteasomes. The antibody (clone MCP165) has been characterised by one- and two dimensional Western blotting. The antibody has not been demonstrated to immunoprecipitate the native 20S proteasome. The immunoglobulin subclass is IgG1.

  • For Immunohistochemistry, Western Blot

Recognizes human and mouse proteasome 20S (subunit beta2). Detects a band of ~29kDa by Western blot.

Type: Primary
Antigen: PSMB2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty