Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PARK2 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
PW9365-0100
89162-346EA 525 USD InStock
89162-346
Anti-PARK2 Rabbit Polyclonal Antibody
Antibodies
Parkin is a ubiquitin-protein ligase (E3), and mutations in the Parkin gene appear to be closely associated with autosomal recessive juvenile Parkinson’s disease. Parkin has been localised to Lewy bodies in patients with sporadic Parkinson’s disease, and in peri-nuclear aggresome-like inclusions in cells over-expressing wild-type Parkin where proteasomal activity has also been inhibited4. Both types of inclusion also contain ubiquitin, α-synuclein, proteasomal subunits and heat shock proteins. A recent paper suggests that loss of proteasomal activity leads to Parkin accumulation with a concomitant reduction in E3 ligase activity resulting in Lewy body formation5.Parkin is selectively recruited from the cytosol to damaged mitochondria to trigger their autophagy (mitophagy).

  • For Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human parkin.

Type: Primary
Antigen: PARK2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty