Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Proteasome 19S Rpt1/S7 Subunit Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
PW9400-0025
89162-360EA 181 USD InStock
89162-360
Anti-Proteasome 19S Rpt1/S7 Subunit Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunoprecipitation

Recognizes human proteasome 26S (19S regulator complex) (Rpt1/S7 subunit).

Type: Primary
Antigen: PROTEASOME 19S RPT1/S7 SUBUNIT
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty