Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SUMO1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
PW9460-0025 PW9460-0100
89162-370EA 525 USD InStock
89162-370 89162-368
Anti-SUMO1 Rabbit Polyclonal Antibody
Antibodies
The small ubiquitin-related modifier SUMO-1 belongs to the growing family of ubiquitin-related proteins involved in post-translational protein modification. It is present in all eukaryotic kingdoms and is highly conserved from yeast to humans. Whereas invertebrates have only one SUMO gene, three members of the SUMO family have been described in vertebrates, SUMO-1 and the close homologues SUMO-2 and SUMO-3 with some 50% homology between SUMO-1 and SUMO-2/3. The SUMO family members have a short N-terminal extension that is absent in ubiquitin, and the function of which is unknown and the sequence of which varies between the three family members.  Unlike ubiquitin, SUMO-1 does not appear to target proteins for degradation but seems to be involved in the modulation of protein-protein interactions. Although having only 18% amino acid sequence identity with ubiquitin, the overall structure closely resembles that of ubiquitin. Whereas the two C-terminal glycine residues required for isopeptide bond formation are conserved between the two molecules, Lys48 found in ubiquitin, and required to generate ubiquitin polymers, is substituted by Gln69in SUMO-1 thereby providing an explanation of why SUMO-1 has not been observed to form polymers.  An increasing number of SUMO-1 substrates are being described but three major substrates for SUMO-1 modification are RanGAP1, PML and IκBα proteins. 

  • For Immunoprecipitation, Western Blot

Recognizes human SUMO-1.

Type: Primary
Antigen: SUMO1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty