Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-G Protein Alpha I3 Subunit Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
BML-SA130-0050
89162-662EA 799 USD InStock
89162-662
Anti-G Protein Alpha I3 Subunit Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunoprecipitation, Western Blot

Recognizes human and cow G protein (alphai3 and alpha0 subunit).

Type: Primary
Antigen: G PROTEIN ALPHA I3 SUBUNIT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Cow (Bovine)

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty