Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NF-KAPPAB P65 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
BML-SA171-0100
89162-698EA 904 USD InStock
89162-698
Anti-NF-KAPPAB P65 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For ELISA, Flow Cytometry, Immunoprecipitation, Western Blot, EMSA

Recognizes human and mouse NFkappaB (p65 subunit).

Type: Primary
Antigen: NF-KAPPAB P65
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty