Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Y Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
BML-SA240-1000
89162-770EA 247 USD InStock
89162-770
Anti-Y Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG2b

  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes phosphotyrosine. Species independent. Does not cross-react with phosphoserine or phosphothreonine.

Type: Primary
Antigen: Y
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Many Species

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty