Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Y Mouse Monoclonal Antibody (Biotin)

Supplier: Enzo Life Sciences
BML-SA243-0100
89162-776EA 358 USD InStock
89162-776
Anti-Y Mouse Monoclonal Antibody (Biotin)
Antibodies
Host: Mouse, Isotype: IgG2b

  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry

Recognizes phosphotyrosine. Species independent. No cross-reactivity with phosphoserine or phosphothreonine.

Type: Primary
Antigen: Y
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Many Species

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty