Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PARP1 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
BML-SA250-0050
89162-782EA 422 USD InStock
89162-782
Anti-PARP1 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For ELISA, Flow Cytometry, Immunohistochemistry, Western Blot

Recognizes an epitope in the C-terminal part of the DNA binding domain of human, mouse, rat, monkey and hamster PARP. Detects a band of ~116kDa by Western blot. Cross-reacts with 85kDa apoptosis-related caspase-3/7-generated cleavage fragment. Cross-reacts with necrosis-related fragments of 50 kDa, 62 kDa and 74 kDa and is therefore useful for distinguishing between these two types of cell death.

Type: Primary
Antigen: PARP1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty