Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PARP Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
BML-SA253-0100 BML-SA253-0025
89162-784EA 186 USD InStock
89162-784 89162-786
Anti-PARP Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For ELISA, Flow Cytometry, Western Blot

Recognizes human, mouse, rat and bovine PARP. Detects a band of ~116kDa by Western blot.

Type: Primary
Antigen: PARP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty