Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-JNK1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
BML-SA272-0020 BML-SA272-0100
89162-814EA 191 USD InStock
89162-814 89162-816
Anti-JNK1 Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunohistochemistry, Western Blot

Recognizes human and rat JNK 1&2 dually phosphorylated at Thr183 and Tyr185. Does not cross-react with unphosphorylated JNK1&2.

Type: Primary
Antigen: JNK1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty