Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EIF2 alpha Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
BML-SA405-0020 BML-SA405-0100
89163-022EA 741 USD InStock
89163-022 89163-020
Anti-EIF2 alpha Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Western Blot

Recognizes human and yeast eIF2alpha phosphorylated at Ser51 and mouse eIF2alpha phosphorylated at Ser52. Does not recognize unphosphorylated form of eIF2alpha.

Type: Primary
Antigen: EIF2 alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty