Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-14-3-3 zeta Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
BML-SA482-0100 BML-SA482-0025
89163-168EA 426.54 USD InStock
89163-168 89163-166
Anti-14-3-3 zeta Rabbit Polyclonal Antibody
Antibodies
Host: Rabbit

  • For Immunohistochemistry, Western Blot, Affinity Chromatography

Recognizes human, mouse, rat, cow, sheep and chicken 14-3-3zeta.

Type: Primary
Antigen: 14-3-3 zeta
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty