Would you like to log in again?
Human Recombinant Casein kinase II (from E. coli)
Supplier: Enzo Life Sciences
BML-SE124-0010
89163-786EA 412.01 USD InStock
89163-786
Human Recombinant Casein kinase II (from E. coli)
Enzymes
Produced in E. coli. Consists of 44kDa alpha catalytic and 26kDa ß regulatory subunits as an alpha2ß2 tetramer.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty