Would you like to log in again?
Human Recombinant Caspase-1 (from E. coli)
Supplier: Enzo Life Sciences
BML-SE168-5000
89163-830EA 633.78 USD InStock
89163-830
Human Recombinant Caspase-1 (from E. coli)
Enzymes
Produced in E. coli. cDNA encodes residues identical to Asn120-His404 (C-terminus) at Genbank Accession No. M87507, except for an Asp381 to Glu change, introduced to stabilize the enzyme against autoproteolysis.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty