Would you like to log in again?
Human Recombinant Caspase-3 (from E. coli)
Supplier: Enzo Life Sciences
BML-SE169-5000
89163-832EA 595.81 USD InStock
89163-832
Human Recombinant Caspase-3 (from E. coli)
Enzymes
Produced in E. coli. The enzyme is cleaved and activated from the proenzyme.
Caspase-3 (CPP32; Yama; apopain) from human cDNA, expressed in an E. coli expression system. The enzyme was cleaved and activated from the proenzyme. Can be used to study enzyme regulation and kinetics, cleave target substrates and screen for inhibitors.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty