Would you like to log in again?
Human Recombinant MMP2 (Catalytic Domain)
Supplier: Enzo Life Sciences
BML-SE237-0010
89163-876EA 583.73 USD InStock
89163-876
Human Recombinant MMP2 (Catalytic Domain)
Proteins and Peptides
Produced in yeast.
Active recombinant matrix metalloproteinase-2 (MMP-2, gelatinase A, 72kDa type IV collagenase) cloned from human cDNA. The enzyme consists of residues Tyr110-Asp452 (NM_004530), which comprises the catalytic/fibronectin domain of human MMP-2, with a C-terminal purification tag. This represents a naturally occurring active form of MMP-2 which lacks the C-terminal hemopexin domain. MMPs lacking this domain cannot cleave native collagens; however, activity toward other targets such as gelatin, casein, or peptide substrates is unaffected.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty