Would you like to log in again?
Human Recombinant Tyrosinase (from E. coli)
Supplier: Enzo Life Sciences
BML-SE535-0100
89164-476EA 467.36 USD InStock
89164-476
Human Recombinant Tyrosinase (from E. coli)
Enzymes
Produced in E. coli.
Tyrosinase is a copper-containing enzyme that catalyzes four steps in the melanin and catecholamine synthesis pathways. Like tyrosine hydroxylase (EC 1.13.16.2), it can catalyze the formation of L-DOPA in the brain and nervous system. It oxidizes monophenols, but like catecholoxidase (EC 1.10.3.1), it can also oxidize o-diphenols to o-quinones. It is involved in cancer, Parkinson’s, pigmentation diseases, Alzheimer’s and obesity. Much of the research and inhibitor screening has used mushroom tyrosinase. However, mushroom tyrosinase interacts very differently with substrates and inhibitors compared to the human enzyme. In addition, commercially available mushroom tyrosinase is not always of good purity. Recombinant versions of human tyrosinase amino acids 1-345 or 1-378, also available commercially, are not functional due the lack of critical catalytic and structural residues.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty