Would you like to log in again?
NEDDylation Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
UW0590-0001
89165-182EA 531.08 USD InStock
89165-182
NEDDylation Kit, Enzo Life Sciences
Protein Linking and Labeling Systems
This kit provides the means of generating a thioester-linked NEDD8-conjugated E2 enzyme, utilizing the first two steps in the NEDD8 cascade, for use in the NEDDylation of E3 ligases and target substrate proteins (user supplied)
  • This kit provides sufficient material for 20 x 20µL reactions
  • Generate NEDD8-E2 thioesters for use in the in vitro NEDDylation of target proteins
  • Demonstrate novel proteins are potential targets for NEDDylation
  • Study NEDD8 activation (E1) and conjugation (E2) steps in the NEDDylation cascade
  • Generate substrates for deNEDDylating enzymes
  • Test proteins for NEDD8 E3 ligase activity
  • Avoid freeze/thaw cycles
Note: This Kit does not contain any NEDD8 E3 ligases.
A NEDD8-specific antibody is provided for detection of NEDDylated proteins via SDS-PAGE and Western blotting.
Delivery information: NEDD8 Activating Enzyme Solution (E1), UbcH12 Solution (E2), NEDD8 Solution NEDDylation Buffer Mg-ATP Solution, NEDD8 Antibody Solution, Non-reducing Gel Loading Buffer.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty