Would you like to log in again?
SUMOylation Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
UW8955-0001
89165-294EA 612.62 USD InStock
89165-294
SUMOylation Kit, Enzo Life Sciences
Protein Linking and Labeling Systems
Kit for generation of SUMOylated proteins in vitro.
  • SUMO-modification of specific proteins in vitro
  • Demonstrate novel proteins are potential targets for SUMOylation under in vitro conditions.
  • Generate substrates for deSUMOylating enzymes, such as SENP1 and SENP2
  • Facilitate or enhance SUMOylation of specific target proteins, particularly under conditions/enzyme concentrations that more closely represent those in vivo
  • SUMOylation of proteins in cell lysates or crude fractions/preparations to facilitate investigation of their role/function in complex solutions
  • Demonstrate SUMOylation of known proteins in specific lysates (confirm with target protein specific antibodies)
  • Use of cell lysate or crude fractions/preparations as source of SUMO E3 ligases to facilitate SUMOylation of purified target proteins in the presence of kit components
This kit provides a means of generating SUMOylated proteins in vitro, by covalent linkage of the carboxy-terminal of SUMO-1, -2 or -3 to specific lysine residues on the target protein via isopeptide bonds, using the SUMOylation enzyme cascade
Suggested uses: For SUMO-modification of specific proteins in vitro, To demonstrate that novel proteins are potential targets for SUMOylation under in vitro conditions, To generate substrates for deSUMOylating enzymes, such as SENP1 and SENP2, To test proteins for SUMO E3 ligase activity.
Note: Protocol provided for application 1. Assay set-up can be readily modified for alternative applications by inclusion, omission or substitution of specific components.
A short sequence containing the consensus ?-K-X-D/E (where lysine is the amino acid modified, ? is a large hydrophobic residue and X is any amino acid residue) is thought to be necessary for this in vitro protein SUMOylation to occur, however SUMOylation has also been observed in cases where the consensus site is not conserved. A control target protein is provided together with all other necessary components. SUMO specific antibodies are provided for detection of SUMOylated proteins via SDS-PAGE and Western blotting. Provides sufficient material for 20 x 20µL reactions.
Delivery information: Kit contains, 20X SUMO Activating Enzyme Solution (SUMO E1) (20 µL), 20X SUMO Conjugating Enzyme Solution (SUMO E2) (20 µL), 20X SUMO Enzyme Solutions (SUMO1, SUMO2, SUMO3) (20 µL each), 10X SUMOylation Buffer (40 µL), 20X Control RanGAP1 SUMOylation Target Protein Solution (RG1) (20 µL), 20X Mg-ATP Solution (20 µL), SUMO1 Antibody Solution (25 µL; SUMO1 (CT) rabbit polyclonal antibody), SUMO2/3 Antibody Solution (25 µL; SUMO2/3 (NT) rabbit polyclonal antibody).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty