Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VIM Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
VG6100-0100
89165-480EA 280 USD InStock
89165-480
Anti-VIM Mouse Monoclonal Antibody
Antibodies
Vimentin is a the type III intermediate filament (IF) protein expressed in a variety of mesenchymal and non-mesenchymal cells, including muscle cells, astrocytes, neurons, endothelial cells and fibroblasts. During development of the mammalian nervous system vimentin is transiently expressed in neuronal precursor cells, often with the type VI IF, nestin. Vimentin appears to play a role in many intracellular processes, recent examples from the literature include adhesion, glial scar formation, neuronal migration, and the normal development of Bergmann glia.

  • For Immunohistochemistry, Western Blot

Recognizes human Vimentin.

Type: Primary
Antigen: VIM
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty