Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ACTA2 Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
ENZ-30933
89165-678EA 285 USD InStock
89165-678
Anti-ACTA2 Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG2b

  • For Immunohistochemistry, Western Blot

Recognizes human, monkey, chicken and rabbit actin isotypes alpha and gamma of smooth muscle cells. Does not cross-react with actin of skeletal or cardiac muscle cells.

Type: Primary
Antigen: ACTA2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty