Would you like to log in again?
BIOARRAY™ Eukaryotic Hybridization Controls, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-42661-150 ENZ-42661-30
89165-780EA 441.9 USD InStock
89165-780 89165-778
BIOARRAY™ Eukaryotic Hybridization Controls, Enzo Life Sciences
Microarray Reagents
Biotin-labeled transcripts for microarray internal controls.
Biotin-labeled transcripts prepared from three E. coli genes, bioB, bioC and bioD, and one from bacteriophage P1, cre. These together with the synthetic biotin-labeled deoxyoligonucleotide control (Deoxyoligo B2) are added to biotin-labeled target cRNA before hybridization to probe sequences on the array. These sequences of known concentration provide an internal control for assaying quality and performance. Deoxyoligo B2 aids software programs such as the Affymetrix Microarray Suite by aligning the grid with the array image.
Delivery information: Includes 20X Eukaryotic Hybridization Controls (30 pM bioB, 100 pM bioC, 500 pM bioD and 2000 pM cre) and Control Deoxyoligo B2 (3 nM).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty