Would you like to log in again?
Bio-16-dUTP, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-42811
89165-828EA 538.82 USD InStock
89165-828
Bio-16-dUTP, Enzo Life Sciences
Nucleic Acid Reagents Nucleotides
Bio-16-dUTP (Biotin-16-2’-deoxyuridine-5’-triphosphate) can replace TTP in reactions in which it serves as a substrate for E. coli DNA polymerase (holoenzyme and Klenow fragment), T4 and Taq DNA polymerases, reverse transcriptase (from AMV and M-MuLV) and terminal transferase.
Bio-16-dUTP can be used to produce biotinylated DNA probes in a variety of labelling reactions including nick translation, random prime labelling, cDNA labelling and 3’-end labelling. The resulting biotin-labeled probe can be used in a variety of hybridization applications incuding Southern blots, Northern blots, or dot blots. The probes can also be used for in situ hybridization procedures on fixed cells and tissues. The biotinylated probes have been shown to hybridize to homologous nucleic acid at the same rate and to the same extent as non-biotinylated probes. The hybridized biotinylated DNA probes can be detected by their interaction with biotin-binding proteins, such as avidin, streptavidin or antibodies coupled to fluorescent dyes or color-producing enzymes.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty