Would you like to log in again?
Red Hydrogen Peroxide Assay Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-51004
89165-870EA 414 USD InStock
89165-870
Red Hydrogen Peroxide Assay Kit, Enzo Life Sciences
Assays Cellular Assays
Red Hydrogen Peroxide Assay Kit provides a simple absorbance-based or fluorometric assay to detect and quantify hydrogen peroxide or peroxidase activity in biochemical assays, cell extracts and in live cells
  • Homogenous assay format, fully compatible with HTS automation
  • Detect as little as 10 picomoles of hydrogen peroxide
  • Mix and read reagents, with minimal hands-on time
It can also be used to detect a variety of oxidase activities through enzyme-coupled reactions. The kit is an optimized ‘mix and read’ assay that is suitable for both 96- and 348-well formats and is compatible with HTS liquid handling.
Delivery information: Kit includes Red Peroxidase Substrate (1 Vial), Horseradish Peroxidase (HRP) (20 units), Hydrogen Peroxide (H2O2) Stock Solution (200 µL), DMSO (1 mL), Assay Buffer (100 mL).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty