Would you like to log in again?
Lyso-ID® Red Detection Kit (GFP-CERTIFIED®), Enzo Life Sciences
Supplier: Enzo Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ENZ-51005-0100 ENZ-51005-500
89165-872EA 345 USD InStock
89165-872 10662-272
Lyso-ID® Red Detection Kit (GFP-CERTIFIED®), Enzo Life Sciences
Assays Cellular Assays
For use with GFP-expressing cell lines, as well as cells expressing blue, cyan or yellow fluorescent proteins.
Lyso-ID® Red Lysosomal Detection Kit (GFP-Certified®) is an ideal choice for fluorescence co-localization imaging with GFP-tagged proteins, providing spectrally pure red signal
The Lyso-ID® Red Lysosomal Detection Kit (GFP-Certified®)contains a proprietary acidic organelle-selective dye suitable for staining live cells. The dye is compatible with most fluorescence detection systems, including conventional and confocal fluorescence microscopes, as well as High Content Screening (HCS) platforms. This kit is specifically designed for use with GFP-expressing cell lines, as well as cells expressing blue, cyan or yellow fluorescent proteins (BFPs, CFPs, YFPs).
The Lyso-ID® Red dye has been validated for utility in live cell imaging applications, showing an appropriate response to treatment with well characterized lysosomal perturbation agents. A lysosome perturbation agent, chloroquine, is provided as a positive control for monitoring changes in lysosome number and volume. A nuclear counterstain (Hoechst 33342) is provided to highlight this organelle as well.
By contrast, many red-emitting lysosomal probes photoconvert to green fluorescent species or display metachromatic artifacts wherein emission occurs both in the red and green regions of the spectrum, leading to spurious results in GFP co-localization experiments.
Delivery information: Kit contains Lyso-ID® Red Detection Reagent, 50 µL, Hoechst 33342 Nuclear Staining, 50 µL, Chloroquine Control, 7.5 µmol and10X Assay Buffer, 15 mL.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty