Would you like to log in again?
TOTAL-NUCLEAR-ID® Green/Red Nucleolar/Nuclear Detection Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-51006-500
89165-874EA 348 USD InStock
89165-874
TOTAL-NUCLEAR-ID® Green/Red Nucleolar/Nuclear Detection Kit, Enzo Life Sciences
Assays Cellular Assays
The kit contains two proprietary dyes suitable for visualizing nucleoli and nuclei simultaneously in living cells.
The dyes allow examination of nucleolar dynamic changes in intracellular distribution, trafficking and localization arising from biological processes such as the cell cycle and ribosome biogenesis
A control nucleolus perturbation agent, actinomycin D, is provided for monitoring changes in nucleolar dynamics. Potential applications for this kit include monitoring impaired ribosome biogenesis, inhibition transcription, cell cycle dynamics and cellular stress, as well as the distribution, trafficking and dynamics of nucleolar proteins, the distribution of viral proteins, and potentially as an aid in identifying cancer cells.
The kit is compatible with most fluorescence detection systems, including conventional and confocal fluorescence microscopes, as well as High Content Screening (HCS) platforms.
Delivery information: Kit contains Nucleolar-ID™ Green Detection Reagent, 50 µL, Nuclear-ID™ Red Detection Reagent, 50 µL, Actinomycin D Control, 125 µg and 10X Assay Buffer, 15 mL.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty