Would you like to log in again?
NUCLEAR-ID® Red Cell Cycle Kit (GFP-CERTIFIED®), Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-51008-100
89165-878EA 488 USD InStock
89165-878
NUCLEAR-ID® Red Cell Cycle Kit (GFP-CERTIFIED®), Enzo Life Sciences
Assays Cellular Assays
Suitable for DNA studies in live, permeabilized and fixed cells for normal cell lines and cell lines exhibiting multiple ploidy levels.
Enzo Life Sciences GFP-Certified™ Nuclear-ID™ Red Cell Cycle Analysis Kit provides a convenient approach for studying the induction and inhibition of cell cycle progression by flow cytometry. The kit is suitable for (1) determining the percentage of cells in a given sample that are in G0/G1, S and G2/M phases, as well as to quantify cells in the sub-G1 phase, and (2) DNA studies in live, permeabilized and fixed cells for normal cell lines and cell lines exhibiting multiple ploidy levels. A control cell cycle perturbation agent, Nocodazole, is provided for monitoring changes in cell cycle dynamics. Potential applications for live-cell studies are in the determination of cellular DNA content and cell cycle distribution for the detection of variations in growth patterns, for monitoring apoptosis, and for evaluating tumor cell behavior and suppressor gene mechanisms.
Ordering information: Quantity: 100 assays Contents: Nuclear-ID™ Red Cell Cycle Detection Reagent, 200 µL
Nocodazole Control, 10 µL
10X Assay Buffer, 15 mL
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty