Would you like to log in again?
NUCLEOLAR-ID® Green Detection Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-51009-500
89165-880EA 254 USD InStock
89165-880
NUCLEOLAR-ID® Green Detection Kit, Enzo Life Sciences
Assays Cellular Assays
Specifically designed for visualizing microscopically nucleoli in living cells.
The kit is compatible with most fluorescence detection systems, including conventional and confocal fluorescence microscopes, as well as High Content Screening (HCS) platforms.
The dye in the kit is resistant to photobleaching, facilitating its use in imaging applications. This nucleolar dye may be used in conjunction with common live-cell nuclear counterstains such as Hoechst 33342, Draq5™, Vybrant® DyeCycle™ Ruby and Enzo’s Nuclear-ID™ Red dye.
A control nucleolus perturbation agent, actinomycin D, is provided for monitoring changes in nucleolar dynamics. Potential applications for this kit include monitoring impaired ribosome biogenesis, inhibition of transcription, cell cycle dynamics and cellular stress, as well as the distribution, trafficking and dynamics of nucleolar proteins, the distribution of viral proteins, and potentially as an aid in identifying cancer cells.
Delivery information: Kit contains Nucleolar-ID™ Green Detection Reagent, 50 µL, Actinomycin D Control, 125 µg and 10X Assay Buffer, 15 mL.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty