Would you like to log in again?
ROS-ID® Total ROS Detection Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-51011
89165-884EA 240 USD InStock
89165-884
ROS-ID® Total ROS Detection Kit, Enzo Life Sciences
Assays Cellular Assays
This kit is designed to directly monitor real time reactive oxygen and/or nitrogen species (ROS/RNS) production in live cells using fluorescence microscopy and/or flow cytometry.
The kit includes Oxidative Stress Detection Reagent (Green) as the major component
The kit is not designed to detect superoxide, reactive chlorine or bromine species, as the fluorescent probe included is relatively insensitive to these analytes. Upon staining, the fluorescent product generated can be visualized using a wide-field fluorescence microscope equipped with standard green filter (490/525 nm), or cytometrically using any flow cytometer equipped with a blue (488 nm) laser.
The Total ROS Detection Kit contains sufficient reagents for at least 200 microscopy assays or 50 flow cytometry assays using live cells (adherent or in solution).
This non-fluorescent, cell-permeable total ROS detection dye reacts directly with a wide range of reactive species, such as hydrogen peroxide, peroxynitrite and hydroxyl radicals, yielding a green fluorescent product indicative of cellular production of different ROS/RNS types.
Delivery information: Kit contains Oxidative Stress Detection Reagent (Green), 300 nmoles, ROS Inducer (Pyocyanin), 1 µmole, ROS Inhibitor (N-acetyl-L-cysteine), 2 x 10 mg and Wash Buffer Salts, 1 pack.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty