Would you like to log in again?
LYSO-ID® Red Cytotoxicity Kit (GFP-CERTIFIED®), Enzo Life Sciences
Supplier: Enzo Life Sciences
51015-KP002
89165-892EA 468 USD InStock
89165-892
LYSO-ID® Red Cytotoxicity Kit (GFP-CERTIFIED®), Enzo Life Sciences
Assays Cellular Assays
A Rapid, Quantitative and HTS‐compatible Live Cell Cytotoxicity Assay
96-well cell-based assay. Provides a rapid and quantitative approach for determining drug- or toxic agent-induced lysosome and lysosome-like organelle perturbations in live cells.
The Lyso-ID® Red Cytotoxicity Kit monitors dysfunction of lysosomal degradation using a drug-like cationic amphiphilic tracer (CAT) dye that rapidly and selectively stains acidic organelles, and is suitable for monitoring accumulation of lysosomes and lysosome-like structures in live cells.
Mechanism of Action
A cationic amphiphilic tracer (CAT) dye that rapidly partitions into cells in a similar manner as drugs that induce phospholipidosis. This tracer was created by placing titratable groups on the probe (dye), to enable labeling to be expanded into lamellar inclusion bodies of cells pretreated with weakly basic cell-permeant compounds, such as the antimalarial drug chloroquine. Besides lysosomes themselves, the probe can be employed for highlighting lysosome-like organelles under conditions wherein cells produce vacuoles that contain most of the degradative enzymes of the lysosome but are not as acidic as the parent organelle.
Ordering information: Quantity: 2 x 96-well plates Contents: 10x dual color detection reagent: 2 x 1ml
Detection buffer: 20ml
Verapamil control: 3μmoles
10x assay buffer: 15ml
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty