Would you like to log in again?
GFP-CERTIFIED® FLUOFORTE® Calcium Assay Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
51020-KP010 51020-KP100
89165-904EA 4460 USD InStock
89165-904 89165-902
GFP-CERTIFIED® FLUOFORTE® Calcium Assay Kit, Enzo Life Sciences
Assays Cellular Assays
The GFP-Certified™ FluoForte™ Calcium Assay kit provides a homogeneous fluorescence-based assay for detecting intracellular calcium mobilization across a broad spectrum of biological targets.
Relative to other commercially available dyes, the GFP-Certified™ FluoForte™ dye is the brightest and most sensitive fluorescent calcium indicator. The kit provides a homogeneous mix-and-read, no-wash calcium mobilization assay. The homogenous cell-based assay for calcium offers fewer steps, lower variability and an easier protocol for adherent and non-adherent cell lines. In addition, it requires neither a washing step, nor exogenous addition of a quencher dye, which could adversely effect receptor-ligand interaction kinetics.
Delivery information: Starter kit includes Reagent A (lyophilized GFP-CERTIFIED® FLUOFORTE® dye), Reagent B (dye efflux inhibitor), Reagent C (Hanks’ buffer with 20mM HEPES).
High-throughput kit includes Reagent A (lyophilized GFP-CERTIFIED® FLUOFORTE® dye), Reagent B (dye efflux inhibitor).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty