Would you like to log in again?
NUCLEAR-ID® Green Chromatin Condensation Detection Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-51021-K200
89165-906EA 406 USD InStock
89165-906
NUCLEAR-ID® Green Chromatin Condensation Detection Kit, Enzo Life Sciences
Assays Cellular Assays
This kit provides a rapid assay for nuclear condensation, a prominent hallmark of apoptosis.A highly permeable green-emitting dye for enhanced detection of apoptosis-induced chromatin condensation.
The Nuclear-ID® Green Chromatin Condensation Detection Kit detects nuclear condensation using a DNA intercalating dye that brightly stains the condensed chromatin of apoptotic cells, but only dimly stains the uncondensed chromatin of healthy living cells.
The cell-permeable dye used in this application is an aromatic, planar cationic structure that inserts between stacked base pairs on the DNA duplex, providing an environmentally-dependent fluorescence enhancement of the dye molecules and large increases in fluorescence signal relative to the free dye in solution. Since the signal enhancement provides a proportional response, direct quantitation of DNA is possible.
Further signal increase is observed upon DNA condensation during apoptosis. Considering the general mutagenic effect of nucleic acid-binding dyes, careful storage and handling of this dye is recommended.
Delivery information: Kit contains Nuclear-ID™ Green Cell Cycle Detection Reagent, 100 µL Apoptosis inducer (Staurosporine), 50nmoles 10X Assay Buffer, 30 mL.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty