Would you like to log in again?
PROTEOSTAT® Thermal Shift Stability Assay Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ENZ-51027-K400 ENZ-51027-K100
89165-918EA 848.65 USD InStock
89165-918 76002-532
PROTEOSTAT® Thermal Shift Stability Assay Kit, Enzo Life Sciences
Assays Protein Assays
This kit has been designed for monitoring protein stability under systematic thermal stress conditions.
  • Accelerated screening for protein stability as a function of pH, ionic strength, and concentration
  • Can detect ligand binding, even without a prior knowledge of the protein’s function or ligand binding site
  • Provides a convenient, complementary orthogonal method for cross-validation of instrumentation-intensive techniques
The aggregation temperature is an indicator of protein stability and can be used to optimize conditions that minimize protein aggregation as well as to identify ligands that bind and confer structural stability to a protein of interest. Conditions that increase the aggregation temperature, increase the stability of the protein.
Delivery information: Kit includes PROTEOSTAT® TS Detection Reagent, β-Lactoglobulin control, 10X Assay Buffer.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty