Would you like to log in again?
Antifade Mounting Medium With Dapi (Hardset), Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-53003-M010
89166-062EA 115 USD InStock
89166-062
Antifade Mounting Medium With Dapi (Hardset), Enzo Life Sciences
Stains Microscopy and staining reagents
Mitochondria dye.
Developed specifically for Mito-ID™ Red mitochondrial stain, but works well with many other stains, such as fluorescein and rhodamine. Causes little or no quenching.
Use of the Mito-ID™ Red Antifade Reagent with DAPI with the Mito-ID™ Red dye, or other stains, significantly extends the photobleaching half-life of the fluorescent stain. The addition of DAPI to the mounting medium allows for staining of the nucleus.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty