Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Melanoma Marker Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
ENZ-C34930
89166-080EA 326 USD InStock
89166-080
Anti-Melanoma Marker Mouse Monoclonal Antibody
Antibodies
Host: Mouse, Isotype: IgG1

  • For Immunohistochemistry

Recognizes a human melanoma-specific antigen present in melanocytic cells and junctional nevi. Does not cross-react with normal melanocytes, intradermal nevus cells or other neoplasms including sarcomas, carcinomas and glial tumors.

Type: Primary
Antigen: MELANOMA MARKER
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty