Would you like to log in again?
Suba·Seal® Septa, Ace Glass
Supplier: Ace Glass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
9107-111 9107-155 9107-143 9107-135 9107-123 9107-142 9107-124 9107-131 9107-130 9107-154 9107-134 9107-110
89166-140PK 362.56 USD InStock
89166-140 89166-130 89166-142 89166-132 89166-144 89166-134 89166-146 89166-136 89166-148 89166-138 89166-128 89166-150
Suba·Seal® Septa, Ace Glass
Stoppers Rubber Stoppers
The Suba·Seal® Septa is a high quality rubber septa designed with serrated bottom for sealing open-top glass vessels as well as standard taper joints
Suba·Seal® Septa have serrated sealing capability to the inside of the vessel, as well as the turn-over top seal. Autoclaveable and packaged in white-room conditions.
Ideal for moisture or air sensitive applications.
Ordering information: Suba-Seal® is a trademark of Sigma-Aldrich Biotechnology LP.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty