Would you like to log in again?
KIMTECH PURE® A5 Clean Processed Apparel, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
12919 88843 88845 12921 88848 88845 88850 88847 88844 12919 88849 88850 88842 12916 88846 12921 88843 88847 88844 88846 12915
89107-050CS 1167.93 USD InStock
89107-050 CA89107-050 89107-040 CA89107-040 89107-042 89107-034 89107-044 CA89107-042 89107-036 89107-046 CA89107-044 CA89107-036 89107-038 89107-048 CA89107-038 89166-196 89166-198 89166-200 89166-202 CA89166-200 CA89166-202
KIMTECH PURE® A5 Clean Processed Apparel, KIMBERLY-CLARK PROFESSIONAL®
Disposable Garments
KIMTECH PURE® A5 Clean Processed apparel offers a high-value combination of comfort and quality, and incoporates innovative design features to provide an exceptionally high standard of protection.
  • High standard of protection and comfort
  • Cloth-like SMS material, contains no natural rubber or latex
  • Manufactured in ISO-certified facility, individually bagged under cleanroom conditions
  • Optional hoods and boot covers designed for use with coveralls
Cloth-like comfort is delivered by the classic Kimberly-Clark SMS (Spunbond Meltblown Spunbond) material that goes into the product
All coveralls features patented REFLEX™ design, which is 7.5 times more rip-resisitant than ANSI minimums. Built-in thumb loops keep sleeves from rolling back and exposing the wearer. Coveralls feature a mandarin collar that rises higher on the neck for added protection. All apparel is equipped with tunneled elastic and bound seams. Triple bagged. All items are suitable for ISO Class 5 or higher cleanrooms.
These garments are manufactured in an ISO certified facility, clean processed, and individually bagged under cleanroom conditions. They contain no natural or rubber latex. The optional hoods and boot covers are designed for use with the coveralls. This A5 apparel is ideally suited for ISO 5–8 Cleanroom environments in pharmaceutical, biotech, and medical device industries. It is easy to don, comfortable to wear, and efficient to stock.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty