Would you like to log in again?
Vacuum Adapter, with PTFE Stopcocks, Chemglass
Supplier: Chemglass
CG-1049-J-29 CG-1049-J-24 CG-1049-J-14 CG-1049-J-30
89166-570EA 256.16 USD InStock
89166-570 89166-572 89166-574 89166-568
Vacuum Adapter, with PTFE Stopcocks, Chemglass
Adapters Adapters, Conical Ground Joints
Vacuum adapter has a ground glass standard taper outer joint on top that provides a convenient leak-free way to connect to disposable long stem filter funnels and a taper inner joint or vial.
  • PTFE stopcock
  • Top outer joint
  • Bottom [ST] inner joint
  • With hose connection
The unique design of the long stem filter funnel and vacuum adapter makes it unnecessary to clean after each use. The 2 mm PTFE stopcock between the outer joint and standard taper inner permits control of the vacuum source. Hose connection is 10 mm OD at the largest serration.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty