Would you like to log in again?
High Recovery Vials, Borosilicate, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CG-4916-35 CG-4916-31 CG-4916-30 CG-4916-32 CG-4916-02 CG-4916-06 CG-4916-34 CG-4916-03 CG-4916-05 CG-4916-33 CG-4916-04 CG-4916-01
89166-728PK 574.95 USD InStock
89166-728 30622-688 89166-718 30622-698 30622-692 89166-720 30622-690 89166-722 30622-696 89166-724 89166-726 30622-694
High Recovery Vials, Borosilicate, Chemglass
Vials
High recovery vial with a conical bottom for complete sample recovery.
  • Boxes are marked with alpha-numeric coordinates for fast and easy vial identification
  • Available complete with open-top caps or vials only
These vials are the same size or slightly smaller on the OD to allow them to be interchanged with any racks or blocks that accept standard dram or scintillation vials. Packed in an SBS coated white tray complete with SBS partition.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty